SY3H3D67FD8E Synthonix Corporation

3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 84545-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47038.html

Safety Information

MSDS https://synthonix.com/47038.html
Availability details for
Quantity
  • Availability
  • Add to Cart