SY3H3D67FD90 Synthonix Corporation

2-Methyl-3-(propyldisulfanyl)furan

  • MDL number No MDL found
  • Cas Number : 61197-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47047.html

Safety Information

MSDS https://synthonix.com/47047.html
Availability details for
Quantity
  • Availability
  • Add to Cart