SY3H3D67FD91 Synthonix Corporation

8-Methyl-8-azabicyclo[3.2.1]oct-3-ene

  • MDL number No MDL found
  • Cas Number : 529-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/47051.html

Safety Information

MSDS https://synthonix.com/47051.html
Availability details for
Quantity
  • Availability
  • Add to Cart