SY3H3D67FD92 Synthonix Corporation

(E)-Methyl 2-((dimethylamino)methylene)-3-oxobutanoate

  • MDL number No MDL found
  • Cas Number : 203186-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47066.html

Safety Information

MSDS https://synthonix.com/47066.html
Availability details for
Quantity
  • Availability
  • Add to Cart