SY3H3D67FD94 Synthonix Corporation

(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

  • MDL number No MDL found
  • Cas Number : 116539-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/47069.html

Safety Information

MSDS https://synthonix.com/47069.html
Availability details for
Quantity
  • Availability
  • Add to Cart