SY3H3D67FD9C Synthonix Corporation

Methyl 5-methyl-3-oxohexanoate

  • Cas Number : 30414-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47097.html

Safety Information

MSDS https://synthonix.com/47097.html
Availability details for
Quantity
  • Availability
  • Add to Cart