SY3H3D67FD9F Synthonix Corporation

Methyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate

  • MDL number No MDL found
  • Cas Number : 135643-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47106.html

Safety Information

MSDS https://synthonix.com/47106.html
Availability details for
Quantity
  • Availability
  • Add to Cart