SY3H3D67FDA5 Synthonix Corporation

(S)-2-((Methoxycarbonyl)amino)-3,3-dimethylbutanoic acid

  • MDL number No MDL found
  • Cas Number : 162537-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47134.html

Safety Information

MSDS https://synthonix.com/47134.html
Availability details for
Quantity
  • Availability
  • Add to Cart