SY3H3D67FDA9 Synthonix Corporation

2-(2-Methoxypropan-2-yl)pyrrolidine

  • MDL number No MDL found
  • Cas Number : 160142-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47173.html

Safety Information

MSDS https://synthonix.com/47173.html
Availability details for
Quantity
  • Availability
  • Add to Cart