SY3H3D67FDAB Synthonix Corporation

(R)-Methyl 2-(isopropylamino)butanoate

  • Cas Number : 947667-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47178.html

Safety Information

MSDS https://synthonix.com/47178.html
Availability details for
Quantity
  • Availability
  • Add to Cart