SY3H3D67FDAD Synthonix Corporation

3-Methoxypyridazine

  • Cas Number : 19064-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41021.html

Safety Information

MSDS https://synthonix.com/41021.html
Availability details for
Quantity
  • Availability
  • Add to Cart