SY3H3D67FDAF Synthonix Corporation

2-Mercaptobenzo[d]thiazole-6-carbonitrile

  • MDL number No MDL found
  • Cas Number : 315228-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/47366.html

Safety Information

MSDS https://synthonix.com/47366.html
Availability details for
Quantity
  • Availability
  • Add to Cart