SY3H3D67FDB1 Synthonix Corporation

Methyl 5-bromo-6-cyanopicolinate

  • MDL number No MDL found
  • Cas Number : 959741-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47569.html

Safety Information

MSDS https://synthonix.com/47569.html
Availability details for
Quantity
  • Availability
  • Add to Cart