SY3H3D67FDB2 Synthonix Corporation

Methyl 6-chloro-5-cyanopicolinate

  • MDL number No MDL found
  • Cas Number : 1254163-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47578.html

Safety Information

MSDS https://synthonix.com/47578.html
Availability details for
Quantity
  • Availability
  • Add to Cart