SY3H3D67FDB6 Synthonix Corporation

Methyl 6-chloro-5-(trifluoromethyl)picolinate

  • MDL number No MDL found
  • Cas Number : 1211518-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/47719.html

Safety Information

MSDS https://synthonix.com/47719.html
Availability details for
Quantity
  • Availability
  • Add to Cart