SY3H3D67FDC1 Synthonix Corporation

Methyl 4-bromo-2-chloro-6-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 1321613-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47773.html

Safety Information

MSDS https://synthonix.com/47773.html
Availability details for
Quantity
  • Availability
  • Add to Cart