SY3H3D67FDC4 Synthonix Corporation

Methyl 4-chloro-2,6-difluorobenzoate

  • Cas Number : 773134-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47782.html

Safety Information

MSDS https://synthonix.com/47782.html
Availability details for
Quantity
  • Availability
  • Add to Cart