SY3H3D67FDC5 Synthonix Corporation

METHYL 2,6-DIFLUORO-3-IODOBENZOATE

  • MDL number No MDL found
  • Cas Number : 501433-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/47785.html

Safety Information

MSDS https://synthonix.com/47785.html
Availability details for
Quantity
  • Availability
  • Add to Cart