SY3H3D67FDD1 Synthonix Corporation

3-Methyl-5-nitrobenzoisothiazole

  • MDL number No MDL found
  • Cas Number : 35272-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47892.html

Safety Information

MSDS https://synthonix.com/47892.html
Availability details for
Quantity
  • Availability
  • Add to Cart