SY3H3D67FDD9 Synthonix Corporation

Methyl 4-bromo-5-cyano-3-methylthiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 648412-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47993.html

Safety Information

MSDS https://synthonix.com/47993.html
Availability details for
Quantity
  • Availability
  • Add to Cart