SY3H3D67FDDD Synthonix Corporation

2-Methoxy-6-(trifluoromethyl)nicotinaldehyde

  • MDL number No MDL found
  • Cas Number : 944904-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/48006.html

Safety Information

MSDS https://synthonix.com/48006.html
Availability details for
Quantity
  • Availability
  • Add to Cart