SY3H3D67FDE0 Synthonix Corporation

3-Methoxy-6-(trifluoromethyl)picolinic acid

  • Cas Number : 1214330-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48028.html

Safety Information

MSDS https://synthonix.com/48028.html
Availability details for
Quantity
  • Availability
  • Add to Cart