SY3H3D67FDE3 Synthonix Corporation

Methyl 3-hydroxy-5-(trifluoromethyl)picolinate

  • MDL number No MDL found
  • Cas Number : 1256810-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48031.html

Safety Information

MSDS https://synthonix.com/48031.html
Availability details for
Quantity
  • Availability
  • Add to Cart