SY3H3D67FDEB Synthonix Corporation

Methyl 2-fluoro-3-nitrobenzoate

  • Cas Number : 946126-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48045.html

Safety Information

MSDS https://synthonix.com/48045.html
Availability details for
Quantity
  • Availability
  • Add to Cart