SY3H3D67FDF0 Synthonix Corporation

Methyl 5-fluoro-2-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 393-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48050.html

Safety Information

MSDS https://synthonix.com/48050.html
Availability details for
Quantity
  • Availability
  • Add to Cart