SY3H3D67FDF5 Synthonix Corporation

Methyl 5-chloro-2-hydroxy-3-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 5043-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48063.html

Safety Information

MSDS https://synthonix.com/48063.html
Availability details for
Quantity
  • Availability
  • Add to Cart