SY3H3D67FDFA Synthonix Corporation

Methyl 2-bromo-5-chlorobenzoate

  • MDL number MFCD00144763
  • Cas Number : 27007-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to pale-yellow to pale-brown powder or crytals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48121.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48121.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart