SY3H3D67FE05 Synthonix Corporation

Methyl 4-bromo-3-fluorobenzoate

  • MDL number MFCD08436044
  • Cas Number : 849758-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48135.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48135.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart