SY3H3D67FE16 Synthonix Corporation

Methyl 3-fluoro-4-iodobenzoate

  • MDL number No MDL found
  • Cas Number : 1027513-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48205.html

Safety Information

MSDS https://synthonix.com/48205.html
Availability details for
Quantity
  • Availability
  • Add to Cart