SY3H3D67FE18 Synthonix Corporation

Methyl thieno[3,4-b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 14630-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48207.html

Safety Information

MSDS https://synthonix.com/48207.html
Availability details for
Quantity
  • Availability
  • Add to Cart