SY3H3D67FE19 Synthonix Corporation

Methyl 3-bromo-5-chloro-2-hydroxybenzoate

  • Cas Number : 4068-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48212.html

Safety Information

MSDS https://synthonix.com/48212.html
Availability details for
Quantity
  • Availability
  • Add to Cart