SY3H3D67FE1A Synthonix Corporation

Methyl 3,5-difluoro-4-hydroxybenzoate

  • Cas Number : 170572-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48228.html

Safety Information

MSDS https://synthonix.com/48228.html
Availability details for
Quantity
  • Availability
  • Add to Cart