SY3H3D67FE1C Synthonix Corporation

1-Methyl-2-(trifluoromethyl)benzene

  • MDL number MFCD00153204
  • Cas Number : 13630-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Almost colorless
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48304.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48304.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart