SY3H3D67FE1F Synthonix Corporation

Methyl 3-amino-4-bromo-2-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 1207175-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48357.html

Safety Information

MSDS https://synthonix.com/48357.html
Availability details for
Quantity
  • Availability
  • Add to Cart