SY3H3D67FE22 Synthonix Corporation

6-Methylpyrido[2,3-b]pyrazine

  • Cas Number : 155629-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48389.html

Safety Information

MSDS https://synthonix.com/48389.html
Availability details for
Quantity
  • Availability
  • Add to Cart