SY3H3D67FE24 Synthonix Corporation

4-Methyl-6-nitro-1H-indazole

  • MDL number No MDL found
  • Cas Number : 885520-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48403.html

Safety Information

MSDS https://synthonix.com/48403.html
Availability details for
Quantity
  • Availability
  • Add to Cart