SY3H3D67FE25 Synthonix Corporation

5-Methyl-6-nitro-1H-indazole

  • Cas Number : 72521-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48404.html

Safety Information

MSDS https://synthonix.com/48404.html
Availability details for
Quantity
  • Availability
  • Add to Cart