SY3H3D67FE26 Synthonix Corporation

7-Methyl-6-nitro-1H-indazole

  • MDL number No MDL found
  • Cas Number : 208457-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48405.html

Safety Information

MSDS https://synthonix.com/48405.html
Availability details for
Quantity
  • Availability
  • Add to Cart