SY3H3D67FE29 Synthonix Corporation

1-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine

  • MDL number No MDL found
  • Cas Number : 1135437-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48410.html

Safety Information

MSDS https://synthonix.com/48410.html
Availability details for
Quantity
  • Availability
  • Add to Cart