SY3H3D67FE2D Synthonix Corporation

6-Methoxy-3-nitro-1H-pyrrolo[3,2-c]pyridine

  • MDL number No MDL found
  • Cas Number : 1190320-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/48417.html

Safety Information

MSDS https://synthonix.com/48417.html
Availability details for
Quantity
  • Availability
  • Add to Cart