SY3H3D67FE2E Synthonix Corporation

6-Methylbenzoxazole

  • Cas Number : 10531-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48438.html

Safety Information

MSDS https://synthonix.com/48438.html
Availability details for
Quantity
  • Availability
  • Add to Cart