SY3H3D67FE30 Synthonix Corporation

5-Methylbenzo[d]isoxazol-3(2H)-one

  • MDL number No MDL found
  • Cas Number : 65685-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48465.html

Safety Information

MSDS https://synthonix.com/48465.html
Availability details for
Quantity
  • Availability
  • Add to Cart