SY3H3D67FE33 Synthonix Corporation

Methyl 5-(dibromomethyl)picolinate

  • MDL number No MDL found
  • Cas Number : 55876-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/48470.html

Safety Information

MSDS https://synthonix.com/48470.html
Availability details for
Quantity
  • Availability
  • Add to Cart