SY3H3D67FE38 Synthonix Corporation

Methyl 2,6-dichloro-4-methylnicotinate

  • MDL number No MDL found
  • Cas Number : 1013648-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48479.html

Safety Information

MSDS https://synthonix.com/48479.html
Availability details for
Quantity
  • Availability
  • Add to Cart