SY3H3D67FE39 Synthonix Corporation

Methyl 4,6-dichloro-2-methylnicotinate

  • MDL number No MDL found
  • Cas Number : 1196073-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48480.html

Safety Information

MSDS https://synthonix.com/48480.html
Availability details for
Quantity
  • Availability
  • Add to Cart