SY3H3D67FE3B Synthonix Corporation

Methyl 2-amino-4-chloro-5-iodobenzoate

  • MDL number No MDL found
  • Cas Number : 199850-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/48483.html

Safety Information

MSDS https://synthonix.com/48483.html
Availability details for
Quantity
  • Availability
  • Add to Cart