SY3H3D67FE3C Synthonix Corporation

Methyl 2-amino-4,5-difluorobenzoate

  • MDL number No MDL found
  • Cas Number : 207346-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48484.html

Safety Information

MSDS https://synthonix.com/48484.html
Availability details for
Quantity
  • Availability
  • Add to Cart