SY3H3D67FE3E Synthonix Corporation

Methyl 4-amino-2,5-difluorobenzoate

  • MDL number No MDL found
  • Cas Number : 952285-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48486.html

Safety Information

MSDS https://synthonix.com/48486.html
Availability details for
Quantity
  • Availability
  • Add to Cart