SY3H3D67FE41 Synthonix Corporation

4-(Methylsulfonyl)benzonitrile

  • MDL number MFCD00216489
  • Cas Number : 22821-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48490.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48490.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart