SY3H3D67FE42 Synthonix Corporation

5-Methoxybenzo[d]thiazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 15193-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48491.html

Safety Information

MSDS https://synthonix.com/48491.html
Availability details for
Quantity
  • Availability
  • Add to Cart